Assay Details
| Gene Symbol: Sntn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503673.2 | XP_003503721.2 | 3 - 4 | 290 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sntn| Gene Symbol: Sntn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503673.2 | XP_003503721.2 | 3 - 4 | 290 | 70 | |
Set Membership: |
|