Assay Details
| Gene Symbol: Cldn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500935.2 | XP_003500983.1 | 4 - 5 | 624 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn15| Gene Symbol: Cldn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500935.2 | XP_003500983.1 | 4 - 5 | 624 | 72 | |
Set Membership: |
|