Assay Details
Gene Symbol: Psors1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505876.2 | XP_003505924.1 | 1 - 2 | 981 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psors1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505876.2 | XP_003505924.1 | 1 - 2 | 981 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |