Assay Details
| Gene Symbol: Tctex1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497384.1 | XP_003497432.1 | 1 | 623 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tctex1d4| Gene Symbol: Tctex1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497384.1 | XP_003497432.1 | 1 | 623 | 59 | |
Set Membership: |
|