Assay Details
| Gene Symbol: Snapin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501662.2 | XP_003501710.2 | 2 - 3 | 209 | 67 | |
| GenBank mRNA | JP065407.1 | - | 2 - 3 | 209 | 67 | |
More Information
Set Membership: |
|

Snapin
Genomic Map