Assay Details
Gene Symbol: Wipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507793.1 | XP_003507841.1 | 7 - 8 | 903 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
