Assay Details
Gene Symbol: Pptc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007627804.1 | XP_007625994.1 | 2 - 3 | 377 | 73 | |
XM_007627811.1 | XP_007626001.1 | 2 - 3 | 200 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
