Assay Details
| Gene Symbol: Mpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511845.2 | XP_003511893.1 | 3 - 4 | 641 | 65 | |
| GenBank mRNA | JI861954.1 | - | 1 - 2 | 180 | 65 | |
More Information
Set Membership: |
|

Mpg
Genomic Map