Assay Details
| Gene Symbol: Vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646545.1 | XP_007644735.1 | 3 - 4 | 448 | 68 | |
| XM_007646550.1 | XP_007644740.1 | 3 - 4 | 448 | 68 | ||
More Information
Set Membership: |
|

Vamp1
Genomic Map