Assay Details
| Gene Symbol: Smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503805.2 | XP_003503853.1 | 2 - 3 | 1908 | 81 | |
| GenBank mRNA | JP045092.1 | - | 2 - 3 | 1129 | 81 | |
More Information
Set Membership: |
|

Smg8
Genomic Map