Assay Details
| Gene Symbol: Ldlrad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503643.2 | XP_003503691.2 | 4 - 5 | 1043 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldlrad2| Gene Symbol: Ldlrad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503643.2 | XP_003503691.2 | 4 - 5 | 1043 | 87 | |
Set Membership: |
|