Assay Details
| Gene Symbol: Wnt5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647078.1 | XP_007645268.1 | 3 - 4 | 673 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt5a| Gene Symbol: Wnt5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647078.1 | XP_007645268.1 | 3 - 4 | 673 | 71 | |
Set Membership: |
|