Assay Details
| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501953.2 | XP_003502001.1 | 5 - 6 | 593 | 87 | |
| XM_007642581.1 | XP_007640771.1 | 4 - 5 | 542 | 87 | ||
More Information
Set Membership: |
|

Serpinb7
Genomic Map