Assay Details
| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504470.2 | XP_003504518.2 | 2 - 3 | 1732 | 82 | |
| XM_003504471.2 | XP_003504519.2 | 2 - 3 | 1379 | 82 | ||
| XM_007645010.1 | XP_007643200.1 | 3 - 4 | 1856 | 82 | ||
| XM_007645011.1 | XP_007643201.1 | 2 - 3 | 1357 | 82 | ||
More Information
Set Membership: |
|

Zbtb26
Genomic Map