Assay Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510262.2 | XP_003510310.1 | 2 - 3 | 736 | 67 | |
| GenBank mRNA | JI868163.1 | - | 1 - 2 | 101 | 67 | |
| JP064160.1 | - | 1 - 2 | 101 | 67 | ||
More Information
Set Membership: |
|

Rilpl2
Genomic Map