Assay Details
| Gene Symbol: Nlrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648871.1 | XP_007647061.1 | 7 - 8 | 2223 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp9| Gene Symbol: Nlrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648871.1 | XP_007647061.1 | 7 - 8 | 2223 | 64 | |
Set Membership: |
|