Assay Details
| Gene Symbol: Csrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647089.1 | XP_007645279.1 | 2 - 3 | 211 | 73 | |
| GenBank mRNA | JI896843.1 | - | 2 - 3 | 192 | 73 | |
| JP047908.1 | - | 2 - 3 | 192 | 73 | ||
More Information
Set Membership: |
|

Csrp2
Genomic Map