Assay Details
| Gene Symbol: Matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498364.1 | XP_003498412.1 | 6 - 7 | 1352 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Matn1| Gene Symbol: Matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498364.1 | XP_003498412.1 | 6 - 7 | 1352 | 76 | |
Set Membership: |
|