Assay Details
| Gene Symbol: Fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499082.2 | XP_003499130.1 | 2 - 3 | 261 | 61 | |
| XM_007638210.1 | XP_007636400.1 | 2 - 3 | 261 | 61 | ||
| GenBank mRNA | JI891039.1 | - | 2 - 3 | 176 | 61 | |
| JP045349.1 | - | 2 - 3 | 176 | 61 | ||
More Information
Set Membership: |
|

Fam32a
Genomic Map