Assay Details
| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509146.2 | XP_003509194.1 | 6 - 7 | 835 | 59 | |
| GenBank mRNA | JI871524.1 | - | 5 - 6 | 440 | 59 | |
| JP058386.1 | - | 5 - 6 | 440 | 59 | ||
More Information
Set Membership: |
|

Timmdc1
Genomic Map