Assay Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507511.2 | XP_003507559.1 | 1 - 2 | 199 | 58 | |
| GenBank mRNA | JI877823.1 | - | 1 - 2 | 182 | 58 | |
| JP060124.1 | - | 1 - 2 | 182 | 58 | ||
More Information
Set Membership: |
|

Tspan31
Genomic Map