Assay Details
| Gene Symbol: Prelid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643883.1 | XP_007642073.1 | 4 - 5 | 286 | 93 | |
| XM_007643884.1 | XP_007642074.1 | 3 - 4 | 209 | 93 | ||
| GenBank mRNA | JI888490.1 | - | 594 | 93 | ||
More Information
Set Membership: |
|

Prelid2
Genomic Map