Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647025.1 | XP_007645215.1 | 2 - 3 | 670 | 112 | |
| GenBank mRNA | JI878202.1 | - | 3 - 4 | 230 | 112 | |
More Information
Set Membership: |
|

Tmem14c
Genomic Map