Assay Details
| Gene Symbol: Hs3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512052.1 | XP_003512100.1 | 2 | 619 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st2| Gene Symbol: Hs3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512052.1 | XP_003512100.1 | 2 | 619 | 73 | |
Set Membership: |
|