Assay Details
| Gene Symbol: Spry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502422.2 | XP_003502470.1 | 2 | 216 | 110 | |
| XM_007643050.1 | XP_007641240.1 | 2 | 125 | 110 | ||
More Information
Set Membership: |
|

Spry4
Genomic Map