Assay Details
| Gene Symbol: Tnfaip8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644059.1 | XP_007642249.1 | 1 | 200 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfaip8| Gene Symbol: Tnfaip8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644059.1 | XP_007642249.1 | 1 | 200 | 115 | |
Set Membership: |
|