Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685543.2 | XP_005685600.1 | 2 - 3 | 425 | 78 | |
| XM_018054395.1 | XP_017909884.1 | 2 - 3 | 195 | 78 | ||
| GenBank mRNA | JO594023.2 | - | 2 - 3 | 159 | 78 | |
More Information
Set Membership: |
|

LRRC57
Genomic Map