Assay Details
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059857.1 | XP_017915346.1 | 6 - 7 | 707 | 64 | |
XM_018059858.1 | XP_017915347.1 | 6 - 7 | 1189 | 64 | ||
XM_018059859.1 | XP_017915348.1 | 6 - 7 | 727 | 64 | ||
XM_018059860.1 | XP_017915349.1 | 6 - 7 | 674 | 64 | ||
XM_018059861.1 | XP_017915350.1 | 6 - 7 | 857 | 64 | ||
GenBank mRNA | JO441265.2 | - | 1 - 2 | 290 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
