Assay Details
| Gene Symbol: LYPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676862.3 | XP_005676919.1 | 5 - 6 | 375 | 62 | |
| GenBank mRNA | JO329401.2 | - | 5 - 6 | 375 | 62 | |
More Information
Set Membership: |
|

LYPLA2
Genomic Map