Assay Details
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285729.1 | NP_001272658.1 | 1023 | 66 | ||
| GenBank mRNA | GQ148781.1 | - | 1023 | 66 | ||
| JO419477.2 | - | 1020 | 66 | |||
More Information
Set Membership: |
|

OTC
Genomic Map