Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041303.1 | XP_017896792.1 | 2 - 3 | 723 | 98 | |
| XM_018041304.1 | XP_017896793.1 | 3 - 4 | 374 | 98 | ||
More Information
Set Membership: |
|

UBTD1
Genomic Map