Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679656.3 | XP_005679713.3 | 1 - 2 | 1019 | 67 | |
| XM_018046722.1 | XP_017902211.1 | 2 - 3 | 523 | 67 | ||
| GenBank mRNA | JO643801.2 | - | 2 - 3 | 539 | 67 | |
More Information
Set Membership: |
|

TMUB1
Genomic Map