Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683019.3 | XP_005683076.1 | 2 - 3 | 323 | 77 | |
| XM_018050306.1 | XP_017905795.1 | 2 - 3 | 324 | 77 | ||
| GenBank mRNA | JO645852.2 | - | 2 - 3 | 285 | 77 | |
| KA338650.1 | - | 6 - 7 | 3992 | 77 | ||
More Information
Set Membership: |
|

DNAJC18
Genomic Map