Assay Details
| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688961.3 | XP_005689018.1 | 1 - 2 | 191 | 76 | |
| GenBank mRNA | KA343475.1 | - | 1 - 2 | 203 | 76 | |
More Information
Set Membership: |
|

SNAI2
Genomic Map