Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700302.3 | XP_005700359.1 | 3 - 4 | 477 | 69 | |
| GenBank mRNA | JT704767.1 | - | 4 - 5 | 443 | 69 | |
More Information
Set Membership: |
|

MOSPD1
Genomic Map