Assay Details
| Gene Symbol: PRELID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682752.3 | XP_005682809.1 | 2 - 3 | 532 | 64 | |
| GenBank mRNA | JO442180.2 | - | 911 | 64 | ||
More Information
Set Membership: |
|

PRELID1
Genomic Map