Assay Details
| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285740.1 | NP_001272669.1 | 6 - 7 | 1239 | 55 | |
| GenBank mRNA | GU586190.1 | - | 6 - 7 | 1239 | 55 | |
| JO419507.2 | - | 4 - 5 | 785 | 55 | ||
More Information
Set Membership: |
|

HMGCS2
Genomic Map