Assay Details
Gene Symbol: FAM107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687957.3 | XP_005688014.1 | 3 - 4 | 651 | 85 | |
XM_018057130.1 | XP_017912619.1 | 2 - 3 | 330 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
