Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061773.1 | XP_017917262.1 | 5 - 6 | 751 | 57 | |
| XM_018061774.1 | XP_017917263.1 | 5 - 6 | 685 | 57 | ||
| GenBank mRNA | JO412150.2 | - | 5 - 6 | 663 | 57 | |
More Information
Set Membership: |
|

OSGIN1
Genomic Map