Assay Details
| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686324.3 | XP_005686381.1 | 2 - 3 | 1103 | 99 | |
| GenBank mRNA | KA346077.1 | - | 1 - 2 | 1280 | 99 | |
More Information
Set Membership: |
|

MRPL53
Genomic Map