Assay Details
| Gene Symbol: SDR39U1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685200.3 | XP_005685257.3 | 5 - 6 | 549 | 84 | |
| XM_018054653.1 | XP_017910142.1 | 4 - 5 | 428 | 84 | ||
| XM_018054654.1 | XP_017910143.1 | 2 - 3 | 224 | 84 | ||
| GenBank mRNA | JO437447.2 | - | 1 - 2 | 744 | 84 | |
| KA320862.1 | - | 1 - 2 | 769 | 84 | ||
More Information
Set Membership: |
|

SDR39U1
Genomic Map