Assay Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692290.3 | XP_005692347.2 | 1 - 2 | 235 | 54 | |
| GenBank mRNA | JO442055.2 | - | 8 - 9 | 771 | 54 | |
| KA319312.1 | - | 1 - 2 | 241 | 54 | ||
More Information
Set Membership: |
|

LIN37
Genomic Map