Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050509.1 | XP_017905998.1 | 6 - 7 | 689 | 91 | |
| XM_018050510.1 | XP_017905999.1 | 6 - 7 | 633 | 91 | ||
| XM_018050511.1 | XP_017906000.1 | 6 - 7 | 584 | 91 | ||
| XM_018050512.1 | XP_017906001.1 | 6 - 7 | 1131 | 91 | ||
| GenBank mRNA | KA304689.1 | - | 3 - 4 | 852 | 91 | |
More Information
Set Membership: |
|

RNF126
Genomic Map