Assay Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063631.1 | XP_017919120.1 | 7 - 8 | 656 | 61 | |
| GenBank mRNA | JO411222.2 | - | 1 - 2 | 899 | 61 | |
| JO433695.2 | - | 7 - 8 | 637 | 61 | ||
| KA325622.1 | - | 1 - 2 | 904 | 61 | ||
More Information
Set Membership: |
|

CHMP6
Genomic Map