Assay Details
| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047067.1 | XP_017902556.1 | 3 - 4 | 566 | 74 | |
| GenBank mRNA | JO377762.2 | - | 3 - 4 | 314 | 74 | |
| JO628743.2 | - | 3 - 4 | 436 | 74 | ||
More Information
Set Membership: |
|

NPY
Genomic Map