Assay Details
| Gene Symbol: SNAPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677488.3 | XP_005677545.1 | 1 - 2 | 220 | 64 | |
| GenBank mRNA | JO644500.2 | - | 1 - 2 | 230 | 64 | |
More Information
Set Membership: |
|

SNAPIN
Genomic Map