Assay Details
| Gene Symbol: TNFAIP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682722.3 | XP_005682779.2 | 1 - 2 | 343 | 107 | |
| GenBank mRNA | JT659444.1 | - | 6 - 7 | 1604 | 107 | |
More Information
Set Membership: |
|

TNFAIP8
Genomic Map