Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976202.2 | XP_013831656.1 | 2 - 3 | 254 | 92 | |
| GenBank mRNA | JO583079.1 | - | 2 - 3 | 430 | 92 | |
| KA316751.1 | - | 2 - 3 | 257 | 92 | ||
More Information
Set Membership: |
|

NXT2
Genomic Map