Assay Details
| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690568.2 | XP_005690625.1 | 3 - 4 | 304 | 88 | |
| GenBank mRNA | KA338263.1 | - | 1 - 2 | 380 | 88 | |
More Information
Set Membership: |
|

MAP1LC3C
Genomic Map