Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060522.1 | XP_017916011.1 | 2 - 3 | 717 | 75 | |
| GenBank mRNA | KA306577.1 | - | 2 - 3 | 1740 | 75 | |
More Information
Set Membership: |
|

VWA1
Genomic Map